1-(2-Chloro-1-methylethyl)pyrrolidine hydrochloride

Supplier: Matrix Scientific

037250-500MG
101842-984EA 217.35 USD
101842-984
1-(2-Chloro-1-methylethyl)pyrrolidine hydrochloride
1-(2-Chloro-1-methylethyl)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₇H₁₅Cl₂N
MDL Number: MFCD12027088
CAS Number: 58414-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR