4-(2-Chlorophenoxy)benzaldehyde
Supplier: Matrix Scientific
037247-500MG
101842-978EA
290.77
USD
101842-978
4-(2-Chlorophenoxy)benzaldehyde
4-(2-Chlorophenoxy)benzaldehyde
Hazard: Irritant
Formula:
C₁₃H₉ClO₂ |
MDL Number:
MFCD07365210 CAS Number: 158771-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...