N-(2-Amino-2-methylpropyl)-N-(3-chlorophenyl)amine
Supplier: Matrix Scientific
037244-500MG
101842-972EA
193.85
USD
101842-972
N-(2-Amino-2-methylpropyl)-N-(3-chlorophenyl)amine
N-(2-Amino-2-methylpropyl)-N-(3-chlorophenyl)amine
Hazard: Irritant
Formula:
C₁₀H₁₅ClN₂ |
MDL Number:
MFCD08691663 CAS Number: 854650-27-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...