4-Chloro-1H-1,2,3-benzotriazol-1-ol

Supplier: Matrix Scientific

037243-500MG
101842-970EA 144.9 USD
101842-970
4-Chloro-1H-1,2,3-benzotriazol-1-ol
4-Chloro-1H-1,2,3-benzotriazol-1-ol

Hazard: Irritant

Formula: C₆H₄ClN₃O
MDL Number: MFCD09028260
CAS Number: 39968-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR