2-(2-Methyl-1H-benzimidazol-1-yl)acetohydrazide

Supplier: Matrix Scientific

037240-500MG
101842-964EA 290.77 USD
101842-964
2-(2-Methyl-1H-benzimidazol-1-yl)acetohydrazide
2-(2-Methyl-1H-benzimidazol-1-yl)acetohydrazide

Hazard: Irritant

Formula: C₁₀H₁₂N₄O
MDL Number: MFCD00227235
CAS Number: 97420-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR