(6-Hydroxypyrimidin-4-yl)acetic acid, sodium salt

Supplier: Matrix Scientific

037235-500MG
101842-954EA 460 USD
101842-954
(6-Hydroxypyrimidin-4-yl)acetic acid, sodium salt
(6-Hydroxypyrimidin-4-yl)acetic acid, sodium salt

Hazard: Irritant

Formula: C₆H₅N₂NaO₃
MDL Number: MFCD28023814
CAS Number: 1119449-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR