[5-(Azepan-1-ylmethyl)-1H-tetrazol-1-yl]acetic acid hydrochloride

Supplier: Matrix Scientific

037230-500MG
101842-944EA 217.35 USD
101842-944
[5-(Azepan-1-ylmethyl)-1H-tetrazol-1-yl]acetic acid hydrochloride
[5-(Azepan-1-ylmethyl)-1H-tetrazol-1-yl]acetic acid hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈ClN₅O₂
MDL Number: MFCD12027082
CAS Number: 1185299-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR