(5-Methyl-1H-tetrazol-1-yl)acetic acid
Supplier: Matrix Scientific
037228-500MG
101842-940EA
335.38
USD
101842-940
(5-Methyl-1H-tetrazol-1-yl)acetic acid
(5-Methyl-1H-tetrazol-1-yl)acetic acid
Hazard: Irritant
Formula:
C₄H₆N₄O₂ |
MDL Number:
MFCD01230441 CAS Number: 21743-55-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...