1-[2-(1H-1,2,4-Triazol-1-yl)phenyl]methanamine hydrochloride

Supplier: Matrix Scientific

Synonyms: 2-(1H-1,2,4-Triazol-1-yl)benzenemethanamine

037227-500MG
101842-938EA 362.25 USD
101842-938
1-[2-(1H-1,2,4-Triazol-1-yl)phenyl]methanamine hydrochloride
1-[2-(1H-1,2,4-Triazol-1-yl)phenyl]methanamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₁ClN₄
MW: 210.67 g/mol
MDL Number: MFCD12027080
CAS Number: 449756-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR