Methyl-4-{[(1-methylethylidene)amino]oxy}benzoate

Supplier: Matrix Scientific

037225-500MG
101842-934EA 146.15 USD
101842-934
Methyl-4-{[(1-methylethylidene)amino]oxy}benzoate
Methyl-4-{[(1-methylethylidene)amino]oxy}benzoate

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD12027078
CAS Number: 58658-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR