(5-Amino-2H-tetrazol-2-yl)acetic acid

Supplier: Matrix Scientific

037224-500MG
101842-932EA 443.08 USD
101842-932
(5-Amino-2H-tetrazol-2-yl)acetic acid
(5-Amino-2H-tetrazol-2-yl)acetic acid

Hazard: Irritant

Formula: C₃H₅N₅O₂
MDL Number: MFCD03419553
CAS Number: 21743-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR