2-(1H-1,2,4-Triazol-1-yl)benzoic acid

Supplier: Matrix Scientific

037222-500MG
101842-928EA 193.85 USD
101842-928
2-(1H-1,2,4-Triazol-1-yl)benzoic acid
2-(1H-1,2,4-Triazol-1-yl)benzoic acid

Hazard: Irritant

Formula: C₉H₇N₃O₂
CAS Number: 138479-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR