2-Chloro-5-(1H-tetrazol-1-yl)benzoic acid

Supplier: Matrix Scientific

037220-500MG
101842-924EA 167.69 USD
101842-924
2-Chloro-5-(1H-tetrazol-1-yl)benzoic acid
2-Chloro-5-(1H-tetrazol-1-yl)benzoic acid

Hazard: Irritant

Formula: C₈H₅ClN₄O₂
MDL Number: MFCD03044411
CAS Number: 190270-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR