6-Methyl-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylic acid
Supplier: Matrix Scientific
037218-500MG
101842-920EA
230.77
USD
101842-920
6-Methyl-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylic acid
6-Methyl-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₈N₂O₃ |
MDL Number:
MFCD01690160 CAS Number: 32092-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...