2-Ethyl-3-methyl-1-benzofuran-7-amine hydrochloride

Supplier: Matrix Scientific

037216-500MG
101842-916EA 246.15 USD
101842-916
2-Ethyl-3-methyl-1-benzofuran-7-amine hydrochloride
2-Ethyl-3-methyl-1-benzofuran-7-amine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄ClNO
MDL Number: MFCD12027075
CAS Number: 174186-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR