3-(2,6-Dioxopiperidin-1-yl)benzoic acid

Supplier: Matrix Scientific

037214-500MG
101842-912EA 387.69 USD
101842-912
3-(2,6-Dioxopiperidin-1-yl)benzoic acid
3-(2,6-Dioxopiperidin-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₁NO₄
MDL Number: MFCD02171888
CAS Number: 140213-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR