2-Phenyl-1-benzofuran-5-amine

Supplier: Matrix Scientific

037213-500MG
101842-910EA 223.08 USD
101842-910
2-Phenyl-1-benzofuran-5-amine
2-Phenyl-1-benzofuran-5-amine

Hazard: Irritant

Formula: C₁₄H₁₁NO
MDL Number: MFCD04971060
CAS Number: 77084-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR