4-[(2-Furylmethyl)thio]aniline
Supplier: Matrix Scientific
037211-500MG
101842-906EA
146.15
USD
101842-906
4-[(2-Furylmethyl)thio]aniline
4-[(2-Furylmethyl)thio]aniline
Hazard: Irritant
Formula:
C₁₁H₁₁NOS |
MDL Number:
MFCD04970973 CAS Number: 869943-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...