3-(4-Fluorophenoxy)-5-nitroaniline

Supplier: Matrix Scientific

037210-500MG
101842-904EA 146.15 USD
101842-904
3-(4-Fluorophenoxy)-5-nitroaniline
3-(4-Fluorophenoxy)-5-nitroaniline

Hazard: Irritant

Formula: C₁₂H₉FN₂O₃
MDL Number: MFCD00731320
CAS Number: 208122-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR