3-[(4-Chlorophenyl)sulfonyl]-5-methoxyaniline

Supplier: Matrix Scientific

037205-500MG
101842-896EA 484.62 USD
101842-896
3-[(4-Chlorophenyl)sulfonyl]-5-methoxyaniline
3-[(4-Chlorophenyl)sulfonyl]-5-methoxyaniline

Hazard: Irritant

Formula: C₁₃H₁₂ClNO₃S
MDL Number: MFCD03419509
CAS Number: 832739-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR