2-Phenyl-1-benzofuran-7-amine

Supplier: Matrix Scientific

037202-500MG
101842-890EA 444.62 USD
101842-890
2-Phenyl-1-benzofuran-7-amine
2-Phenyl-1-benzofuran-7-amine

Hazard: Irritant

Formula: C₁₄H₁₁NO
MDL Number: MFCD04971061
CAS Number: 77083-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR