Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-2-ethyl-3-methyl-1-benzofuran-7-amine hydrochloride

Supplier: Matrix Scientific


037200-500MG
101842-886EA 247.15 USD InStock
101842-886
5-Chloro-2-ethyl-3-methyl-1-benzofuran-7-amine hydrochloride
5-Chloro-2-ethyl-3-methyl-1-benzofuran-7-amine hydrochloride
CAS: 1185301-77-2
MDL Number: MFCD12027073
Molecular Formula: C11H13Cl2NO
Molecular Weight: 246,14
Hazard: Irritant


Formula: C₁₁H₁₃Cl₂NO
MW: 246,14 g/mol
MDL Number: MFCD12027073
CAS Number: 1185301-77-2
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty