3-Nitro-5-[3-(trifluoromethyl)phenoxy]aniline

Supplier: Matrix Scientific

037198-500MG
101842-882EA 144.9 USD
101842-882
3-Nitro-5-[3-(trifluoromethyl)phenoxy]aniline
3-Nitro-5-[3-(trifluoromethyl)phenoxy]aniline

Hazard: Irritant

Formula: C₁₃H₉F₃N₂O₃
MDL Number: MFCD04969805
CAS Number: 832741-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR