3-(2,4-Difluorophenoxy)-5-nitroaniline

Supplier: Matrix Scientific

037197-500MG
101842-880EA 109.25 USD
101842-880
3-(2,4-Difluorophenoxy)-5-nitroaniline
3-(2,4-Difluorophenoxy)-5-nitroaniline

Hazard: Irritant

Formula: C₁₂H₈F₂N₂O₃
MDL Number: MFCD02089312
CAS Number: 310451-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR