1-[2-(2-Fluorophenoxy)phenyl]-1H-tetrazole

Supplier: Matrix Scientific

037193-500MG
101842-872EA 109.25 USD
101842-872
1-[2-(2-Fluorophenoxy)phenyl]-1H-tetrazole
1-[2-(2-Fluorophenoxy)phenyl]-1H-tetrazole

Hazard: Irritant

Formula: C₁₃H₉FN₄O
MDL Number: MFCD09064561
CAS Number: 924823-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR