N-5-Phenyl-1H-tetrazole-1,5-diamine

Supplier: Matrix Scientific

037190-500MG
101842-866EA 109.25 USD
101842-866
N-5-Phenyl-1H-tetrazole-1,5-diamine
N-5-Phenyl-1H-tetrazole-1,5-diamine

Hazard: Irritant

Formula: C₇H₈N₆
MDL Number: MFCD00468279
CAS Number: 5533-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR