2-Amino-N-phenylethanesulfonamide hydrochloride

Supplier: Matrix Scientific

037182-500MG
101842-850EA 444.62 USD
101842-850
2-Amino-N-phenylethanesulfonamide hydrochloride
2-Amino-N-phenylethanesulfonamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₃ClN₂O₂S
MDL Number: MFCD11858079
CAS Number: 1100424-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR