1-Bromo-4-isopropoxybenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-4-isopropoxylbenzene

037177-5G
101842-838EA 207.69 USD
101842-838
1-Bromo-4-isopropoxybenzene ≥97%
1-Bromo-4-isopropoxybenzene

Hazard: Irritant

Formula: C₉H₁₁BrO
MW: 215.1 g/mol
Boiling Pt: 119…120 °C (20 Torr)
Melting Pt: 17 °C
MDL Number: MFCD00089972
CAS Number: 6967-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR