2-(Piperazin-1-ylsulfonyl)ethanamine oxalate

Supplier: Matrix Scientific

037168-500MG
101842-816EA 193.85 USD
101842-816
2-(Piperazin-1-ylsulfonyl)ethanamine oxalate
2-(Piperazin-1-ylsulfonyl)ethanamine oxalate

Hazard: Irritant

Formula: C₈H₁₇N₃O₆S
MDL Number: MFCD12027067
CAS Number: 1185299-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR