Ethyl-2-chloroquinoline-4-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 2-chloroquinoline-4-carboxylate

037160-500MG
101842-800EA 193.85 USD
101842-800
Ethyl-2-chloroquinoline-4-carboxylate
Ethyl 2-chloroquinoline-4-carboxylate

Hazard: Irritant

Formula: C₁₁H₈ClNO₂
MW: 221.6397 g/mol
MDL Number: MFCD03413689
CAS Number: 62482-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR