7-Amino[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylic acid

Supplier: Matrix Scientific

037156-500MG
101842-792EA 113.85 USD
101842-792
7-Amino[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylic acid
7-Amino[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylic acid

Hazard: Irritant

Formula: C₆H₅N₅O₂
MDL Number: MFCD11058063
CAS Number: 89488-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR