Ethyl-1-methyl-4-oxopiperidine-3-carboxylateoxalate

Supplier: Matrix Scientific

037155-500MG
101842-790EA 193.85 USD
101842-790
Ethyl-1-methyl-4-oxopiperidine-3-carboxylateoxalate
Ethyl-1-methyl-4-oxopiperidine-3-carboxylateoxalate

Hazard: Irritant

Formula: C₁₁H₁₇NO₇
MDL Number: MFCD12027063
CAS Number: 37673-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR