2-(2-Chloroethyl)-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

Synonyms: 2-(2-Chloroethyl)isoindoline-1,3-dione

037154-500MG
101842-788EA 193.85 USD
101842-788
2-(2-Chloroethyl)-1H-isoindole-1,3(2H)-dione
2-(2-Chloroethyl)-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₀H₈ClNO₂
MW: 209.63 g/mol
MDL Number: MFCD00129356
CAS Number: 6270-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR