Ethyl-4-methyl-2-(methylamino)-1,3-thiazole-5-carboxylate

Supplier: Matrix Scientific

037153-500MG
101842-786EA 59.8 USD
101842-786
Ethyl-4-methyl-2-(methylamino)-1,3-thiazole-5-carboxylate
Ethyl-4-methyl-2-(methylamino)-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₈H₁₂N₂O₂S
MDL Number: MFCD07269200
CAS Number: 3161-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR