4-Methyl-6-nitroquinolin-2-ol
Supplier: Matrix Scientific
037152-500MG
101842-784EA
221.54
USD
101842-784
4-Methyl-6-nitroquinolin-2-ol
4-Methyl-6-nitroquinolin-2-ol
Hazard: Irritant
Formula:
C₁₀H₈N₂O₃ |
MDL Number:
MFCD01014320 CAS Number: 90771-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...