N-[1-(4-Bromophenyl)ethyl]-N,N-dimethylamine hydrochloride
Supplier: Matrix Scientific
037148-500MG
101842-778EA
193.85
USD
101842-778
N-[1-(4-Bromophenyl)ethyl]-N,N-dimethylamine hydrochloride
N-[1-(4-Bromophenyl)ethyl]-N,N-dimethylamine hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₅BrClN |
MDL Number:
MFCD12027060 CAS Number: 140621-51-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...