Ethyl-3-hydroxyquinoxaline-2-carboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 3-Oxo-3,4-dihydro-2-quinoxalinecarboxylate

037146-500MG
101842-774EA 144.9 USD
101842-774
Ethyl-3-hydroxyquinoxaline-2-carboxylate
Ethyl-3-hydroxyquinoxaline-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₃
MW: 218.21 g/mol
MDL Number: MFCD00054595
CAS Number: 36818-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR