N,N-Bis(2-isopropoxyethyl)amine

Supplier: Matrix Scientific
Warning

Synonyms: Bis(2-isopropoxyethyl)amine

037145-500MG
101842-772EA 144.9 USD
101842-772
N,N-Bis(2-isopropoxyethyl)amine
N,N-Bis(2-isopropoxyethyl)amine
Formula: C₁₀H₂₃NO₂
MW: 189.3 g/mol
MDL Number: MFCD06797066
CAS Number: 1119451-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR