5-Methyl-7-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

Supplier: Matrix Scientific

037144-500MG
101842-770EA 387.69 USD
101842-770
5-Methyl-7-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidin-2-amine
5-Methyl-7-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

Hazard: Irritant

Formula: C₇H₆F₃N₅
MW: 217.15 g/mol
MDL Number: MFCD00472946
CAS Number: 339201-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR