(4Z)-1,5,6,7-Tetrahydro-4H-indazol-4-one oxime
Supplier: Matrix Scientific
037141-500MG
101842-764EA
193.85
USD
101842-764
(4Z)-1,5,6,7-Tetrahydro-4H-indazol-4-one oxime
(4Z)-1,5,6,7-Tetrahydro-4H-indazol-4-one oxime
Hazard: Irritant
Formula:
C₇H₉N₃O |
MDL Number:
MFCD12027058 CAS Number: 1119451-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...