6-Bromo-2,3,4,9-tetrahydro-1H-carbazole

Supplier: Matrix Scientific

037140-500MG
101842-762EA 37.95 USD
101842-762
6-Bromo-2,3,4,9-tetrahydro-1H-carbazole
6-Bromo-2,3,4,9-tetrahydro-1H-carbazole

Hazard: Irritant

Formula: C₁₂H₁₂BrN
MDL Number: MFCD00094989
CAS Number: 21865-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR