4-Methyl-1H-1,2,3-benzotriazole

Supplier: Matrix Scientific

037135-500MG
101842-750EA 193.85 USD
101842-750
4-Methyl-1H-1,2,3-benzotriazole
4-Methyl-1H-1,2,3-benzotriazole

Hazard: Irritant

Formula: C₇H₇N₃
MW: 133.15 g/mol
MDL Number: MFCD00035800
CAS Number: 29878-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR