N-(5-Amino-1H-1,2,4-triazol-3-yl)acetamide

Supplier: Matrix Scientific

037132-500MG
101842-744EA 461.54 USD
101842-744
N-(5-Amino-1H-1,2,4-triazol-3-yl)acetamide
N-(5-Amino-1H-1,2,4-triazol-3-yl)acetamide

Hazard: Irritant

Formula: C₄H₇N₅O
MDL Number: MFCD01230436
CAS Number: 80616-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR