4-Methyl-6-(trifluoromethyl)pyrimidin-2-ol
Supplier: Matrix Scientific
037129-500MG
101842-740EA
193.85
USD
101842-740
4-Methyl-6-(trifluoromethyl)pyrimidin-2-ol
4-Methyl-6-(trifluoromethyl)pyrimidin-2-ol
Hazard: Irritant
Formula:
C₆H₅F₃N₂O |
MDL Number:
MFCD06617941 CAS Number: 91606-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...