[3-(4-Methoxyphenyl)-1H-pyrazol-1-yl]acetic acid

Supplier: Matrix Scientific

037128-500MG
101842-738EA 290.77 USD
101842-738
[3-(4-Methoxyphenyl)-1H-pyrazol-1-yl]acetic acid
[3-(4-Methoxyphenyl)-1H-pyrazol-1-yl]acetic acid

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₃
MDL Number: MFCD00665860
CAS Number: 959584-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR