[1-(4-Fluorophenyl)-1H-pyrazol-4-yl]methanol

Supplier: Matrix Scientific

037126-500MG
101842-734EA 217.35 USD
101842-734
[1-(4-Fluorophenyl)-1H-pyrazol-4-yl]methanol
[1-(4-Fluorophenyl)-1H-pyrazol-4-yl]methanol

Hazard: Irritant

Formula: C₁₀H₉FN₂O
MDL Number: MFCD08059807
CAS Number: 153863-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR