3-Aminoquinoxaline-2-carbonitrile 1,4-dioxide

Supplier: Matrix Scientific

037125-500MG
101842-732EA 116.92 USD
101842-732
3-Aminoquinoxaline-2-carbonitrile 1,4-dioxide
3-Aminoquinoxaline-2-carbonitrile 1,4-dioxide

Hazard: Irritant

Formula: C₉H₆N₄O₂
MDL Number: MFCD00102307
CAS Number: 23190-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR