3-(4-Aminophenyl)-1,2,3,4-oxatriazol-3-ium-5-olate

Supplier: Matrix Scientific

037124-500MG
101842-730EA 387.69 USD
101842-730
3-(4-Aminophenyl)-1,2,3,4-oxatriazol-3-ium-5-olate
3-(4-Aminophenyl)-1,2,3,4-oxatriazol-3-ium-5-olate

Hazard: Irritant

Formula: C₇H₆N₄O₂
MDL Number: MFCD12027055
CAS Number: 1263143-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR