1-(3,4-Dimethylphenyl)-1H-pyrazole-4-carbaldehyde

Supplier: Matrix Scientific

037123-500MG
101842-728EA 146.15 USD
101842-728
1-(3,4-Dimethylphenyl)-1H-pyrazole-4-carbaldehyde
1-(3,4-Dimethylphenyl)-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₂N₂O
MDL Number: MFCD09864595
CAS Number: 1171494-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR