4-Methyl-2-phenylpyrimidine-5-carboxylic acid

Supplier: Matrix Scientific

037117-500MG
101842-714EA 217.35 USD
101842-714
4-Methyl-2-phenylpyrimidine-5-carboxylic acid
4-Methyl-2-phenylpyrimidine-5-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₂
MDL Number: MFCD02682018
CAS Number: 103249-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR